Skip to Content
Merck
All Photos(1)

Key Documents

567750

Sigma-Aldrich

Splitomicin

A cell-permeable lactone derived from β-naphthol that acts as a selective inhibitor of NAD+-dependent histone deacetylase activity of Sir2 protein (IC₅₀ = 60 µM).

Synonym(s):

Splitomicin, 1,2-Dihydro-3H-naphtho[2,1-b]pyran-3-one

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€190.00

€190.00


Estimated to ship on05 May 2025Details



Select a Size

Change View
5 MG
€190.00

About This Item

Empirical Formula (Hill Notation):
C13H10O2
CAS Number:
Molecular Weight:
198.22
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€190.00


Estimated to ship on05 May 2025Details


Quality Level

Assay

≥95% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

O1c2c(c3c(cc2)cccc3)CCC1=O

InChI

1S/C13H10O2/c14-13-8-6-11-10-4-2-1-3-9(10)5-7-12(11)15-13/h1-5,7H,6,8H2

InChI key

ISFPDBUKMJDAJH-UHFFFAOYSA-N

General description

A cell-permeable and selective inhibitor of NAD+-dependent deacetylase activity of Sir2 protein (IC50 = 60 µM). Creates a phenocopy of the sir2 deletion mutant in S. cerevisiae and sensitizes mammalian cells to a variety of DNA-damaging agents by abrogating Sir2p activity on p53. Reported to act by either altering or blocking access to the acetylated histone-binding pocket.
A cell-permeable lactone derived from β-naphthol that acts as a selective inhibitor of NAD+-dependent histone deacetylase activity of Sir2 protein (IC50 = 60 µM). It creates a conditional phenocopy of a sir2 deletion mutant in S. cerevisiae and sensitizes mammalian cells to a variety of DNA-damaging agents by abrogating Sir2p activity on p53. Acts by either altering or blocking access to the acetylated histone binding pocket.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
NAD+-dependent histone deacetylase activity of Sir2 protein
Product does not compete with ATP.
Reversible: no
Target IC50: 60 µM against NAD+-dependent histone deacetylase activity of Sir2 protein

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Bedalov, A., et al. 2001. Proc. Natl. Acad. Sci. USA98, 15113.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service