Skip to Content
Merck
All Photos(2)

Key Documents

56755-M

Millipore

Sorbitol

A sugar alcohol produced by reduction of glucose.

Synonym(s):

D-Sorbitol, D-Glucitol, D-Glucitol

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
€81.00

€81.00


Estimated to ship on08 April 2025Details


Request a Bulk Order

Select a Size

Change View
1 KG
€81.00

About This Item

Empirical Formula (Hill Notation):
C6H14O6
CAS Number:
Molecular Weight:
182.17
Beilstein:
1721899
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25

€81.00


Estimated to ship on08 April 2025Details


Request a Bulk Order

vapor density

<1 (vs air)

Quality Level

vapor pressure

<0.1 mmHg ( 25 °C)

description

Merck USA index - 14, 8725

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

mp

98-100 °C (lit.)

solubility

water: 400 mg/mL

storage temp.

10-30°C

SMILES string

OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)CO

InChI

1S/C6H14O6/c7-1-3(9)5(11)6(12)4(10)2-8/h3-12H,1-2H2/t3-,4+,5-,6-/m1/s1

InChI key

FBPFZTCFMRRESA-JGWLITMVSA-N

Looking for similar products? Visit Product Comparison Guide

General description

A sugar alcohol produced by reduction of glucose.

Application


  • Molecular Correlates of Diapause in Aphidoletes aphidimyza.: This study explores the molecular basis of diapause in the predatory midge Aphidoletes aphidimyza, highlighting the role of sorbitol in stress resistance and energy storage during diapause (Dai et al., 2024).

  • Sugar substitutes on caries prevention in permanent teeth among children and adolescents: a systematic review and meta-analysis.: This review examines the efficacy of sugar substitutes, including sorbitol, in preventing dental caries, showing significant benefits in caries reduction (Luo et al., 2024).

  • Membrane Permeability Dominates over Electrostatic Interactions in Dictating Drug Transport in Osmotically Shocked Escherichia coli.: Investigates the role of sorbitol-induced osmotic shock in altering membrane permeability, which impacts drug transport in Escherichia coli, emphasizing the significance of sorbitol in biophysical studies of bacterial membranes (Kumar et al., 2024).

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, store at room temperature (20°C). Stock solutions are stable for up to 6 months at 20°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service