Skip to Content
Merck
All Photos(1)

Key Documents

567430

Sigma-Aldrich

Sodium Butyrate

A short chain fatty acid that has been shown to induce apoptosis in colon carcinoma cell lines by a p53-independent pathway.

Synonym(s):

Sodium Butyrate, Butyric Acid, Na, NaB, HDAC Inhibitor V

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€84.70

€84.70


Estimated to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
250 MG
€84.70

About This Item

Empirical Formula (Hill Notation):
C4H7NaO2
CAS Number:
Molecular Weight:
110.09
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€84.70


Estimated to ship on22 April 2025Details


Request a Bulk Order

Quality Level

description

RTECS - ET6400000

Assay

≥98% (titration)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white

solubility

water: 2 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

[Na+].[O-]C(=O)CCC

InChI

1S/C4H8O2.Na/c1-2-3-4(5)6;/h2-3H2,1H3,(H,5,6);/q;+1/p-1

InChI key

MFBOGIVSZKQAPD-UHFFFAOYSA-M

General description

A short chain fatty acid that has been shown to induce apoptosis in colon carcinoma cell lines by a p53-independent pathway. Inhibits c-myc splicing and interferes with signal transduction processes, including the release of Ca2+ from intracellular stores. Reduces histamine-induced calcium mobilization in C6 glioma cells. Down-regulates the activity of the serine/threonine protein kinase casein kinase II in HT29 cells.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Induce apoptosis
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Ou, H.C., et al. 1997. Neuroreport 8, 1375.
Russo, G.L., et al. 1997. Biochem. Biophys. Res. Commun. 233, 673.
Krupitza, G., et al. 1995. Carcinogenesis16, 1199.
Calabresse, C., et al. 1993. Biochem. Biophys. Res. Commun.195, 31.
Hague, A., et al. 1993. Int. J. Cancer55, 498.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

C Pellegrini et al.
NPJ Parkinson's disease, 8(1), 9-9 (2022-01-14)
Bowel inflammation, impaired intestinal epithelial barrier (IEB), and gut dysbiosis could represent early events in Parkinson's disease (PD). This study examined, in a descriptive manner, the correlation among enteric α-synuclein, bowel inflammation, impairments of IEB and alterations of enteric bacteria

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service