Skip to Content
Merck
All Photos(1)

Key Documents

566328

Sigma-Aldrich

HDAC Activity Assay Kit

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
€488.00

€488.00


Estimated to ship on02 May 2025Details



Select a Size

Change View
1 KIT
€488.00

About This Item

UNSPSC Code:
41116158
NACRES:
NA.84

€488.00


Estimated to ship on02 May 2025Details


Quality Level

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Resp. Sens. 1 - Skin Irrit. 2

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service