Skip to Content
Merck
All Photos(1)

Key Documents

55714

Millipore

Ribonucleic Acid, Yeast

Synonym(s):

Ribonucleic Acid, Yeast

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€292.00
1 G
€999.00

€292.00


In StockDetails


Request a Bulk Order

Select a Size

Change View
250 MG
€292.00
1 G
€999.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352208
NACRES:
NA.51

€292.00


In StockDetails


Request a Bulk Order

form

solid

Quality Level

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white to off-white

solubility

soluble (1N NaOH (5%))

shipped in

ambient

storage temp.

−20°C

General description

Polynucleotide involved in protein synthesis. Found in both the nucleus and the cytoplasm of cells.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C).

Other Notes

Holley, R.W., et al. 1961. J. Biol. Chem.236, 200.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service