Skip to Content
Merck
All Photos(1)

Key Documents

555562

Sigma-Aldrich

Rho Kinase Inhibitor XIII, RKI-1447

Synonym(s):

Rho Kinase Inhibitor XIII, RKI-1447, 1-(3-Hydroxybenzyl)-3-(4-(pyridin-4-yl)thiazol-2-yl)urea, Methanesulfonate, ROCK Inhibitor XIII, 1-(3-Hydroxybenzyl)-3-(4-(pyridin-4-yl)thiazol-2-yl)urea, Mesylate

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€245.00

€245.00


Estimated to ship on31 March 2026Details



Select a Size

Change View
10 MG
€245.00

About This Item

Empirical Formula (Hill Notation):
C16H14N4O2S · xCH4O3S
Molecular Weight:
326.37 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77

€245.00


Estimated to ship on31 March 2026Details


Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

yellow-white

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A cell-permeable pyridylthiazolyl-urea that acts as a potent, ATP site-targeting, Rho Kinase inhibitor (IC50 = 14.5 and 6.2 nM against ROCK1 and ROCK2, respectively), displaying much reduced potency against PKA, PKN1/PRK1, p70S6K/RPS6kB1, AKT1, MRCKa/CDC42BPA (85.5%, 80.5%, 61.9%, 56.0%, and 50.4% inhibition, respectively, at 1 µM) and little or no activity against 15 other kinases (IC50 >1 µM). Selectively inhibits ROCK-dependent MLC-2 Ser19 and MYPT-1 Thr696 phosphorylations (effective conc. 0.1 to 1 µM in MDA-MB-231, MDA-MB-468, and H1299 cultures), but not mTORC2-dependent Akt Ser473, PAK-dependent Mek Ser298, or S6K-dependent S6 Ser240/244 phosphorylations (up to 10 µM in MDA-MB-231 cells). Shown to inhibit MDA-MB-231 invasion (by 53% and 85%, respectively, with 1 and 10 µM inhibitor) and anchorage-independent colony formation (IC50 = 709 nM) in vitro and be efficacious in suppressing established tumor expansion in a murine ErbB2 mammary cancer model (8.8% vs. 68.3% average tumor size expansion in 14 days, respectively, with or without treatment; 14 dailly i.p. doses at 200 mg/kg) in vivo.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Rho kinases 1 & 2
Reversible: yes
Target IC50: 14.5 and 6.2 nM against ROCK1 and ROCK2, respectively

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Patel, R.A., et al. 2012. Cancer Res.72, 5025.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service