Skip to Content
Merck
All Photos(1)

Documents

540216

Sigma-Aldrich

PTP Inhibitor XVIII

The PTP Inhibitor XVIII, also referenced under CAS 1229246-07-4, controls the biological activity of PTP. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

PTP Inhibitor XVIII, Protein Tyrosine Phosphatase Inhibitor XIII, 5-( E)-(2-(3,5-Ditrifluoromethylbenzyloxy)-5-bromo)-benzylidene-1,3-thiazolane-2,4-dione, PPAR Activator VII, Protein Tyrosine Phosphatase Inhibitor XIII, 5-(E)-(2-(3,5-Ditrifluoromethylbenzyloxy)-5-bromo)-benzylidene-1,3-thiazolane-2,4-dione, PPAR Activator VII

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H10BrF6NO3S
CAS Number:
Molecular Weight:
526.25
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

light yellow

solubility

DMSO: 25 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A cell-permeable thiazolidinedione compound that is shown to inhibit the enzymatic activity of PTP1B, SHP-1, YOP, TC-PTP, and YPTP1 (IC50 = 5, 5.8, 6, 16, and 18 µM, respectively), but not that of LAR even at concentrations as high as 100 µM. Also reported to activate cellular PPAR with an comparable potency seen with troglitazone (Cat. No. 648469). The combined effect of PTP1B inhibition and PPAR activation most likely accounts for the compound′s in vivo antiobesity and antihypoglycemic activity when supplemented directly into the daily high fat diet of C57BL/6j jms Slc male mice (2 mg/g food, ~143 mg/kg animal weight or 4.8 mg/animal).

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Bhattarai, B.R., et al. 2009. Bioorg. Med. Chem. Lett.19, 6161.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service