Skip to Content
Merck
All Photos(1)

Key Documents

539722

Millipore

ProteoExtract® Phosphopeptide Enrichment TiO₂ Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106500
NACRES:
NA.77

usage

sufficient for 100 isolation(s)

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

technique(s)

mass spectrometry (MS): suitable

shipped in

wet ice

storage temp.

2-8°C

General description

The ProteoExtract Phosphopeptide TiO2 Enrichment Kit is a useful tool for enrichment of phosphorylated peptides from complex samples by a highly selective Titanium oxide solid phase. It enhances the ability to routinely identify and characterize large numbers of phosphorylated species within complex protein mixtures taking advantage of a novel titanium dioxide material. Captured and eluted phosphopeptide fractions are ready for MALDI- and ESI- mass spectrometry analysis.

Components

TiO₂ Phosphobind Resin, 10X TiO₂ Phosphobind Buffer, Wash Buffer 1, Wash Buffer 2, TiO₂ Elution Buffer, Dihydroxybenzoic Acid, and a user protocol.

Warning

Toxicity: Multiple Toxicity Values, refer to MSDS (O)

Principle

The ProteoExtract Phosphopeptide Enrichment TiO₂ Kit is designed to enrich phosphorylated peptides from complex samples using a solid phase, highly selective, titanium oxide. The enriched phosphopeptide fraction is directly compatible with downstream analysis by MALDI and ESI mass spectrometry.

Storage and Stability

Upon arrival store the entire contents of the kit at 4°C. Following reconstitution of the Dihydroxybenzoic Acid refrigerate (4°C) for short-term storage or aliquot and freeze (-20°C) for long-term storage; reconstituted Dihydroxybenzoid Acid is stable for up to 6 weeks at 4°C or for up to 6 months at -20°C.

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.

Legal Information

Not available for sale or import into the United States
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
PROTEOEXTRACT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

3 - Flammable Liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service