Skip to Content
Merck
All Photos(1)

Key Documents

539690

Sigma-Aldrich

Protein Synthesis Initiation Inhibitor, NSC 119889

The Protein Synthesis Initiation Inhibitor, NSC 119889 blocks 5′-mediated/cap-dependent initiation of protein synthesis. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Synonym(s):

Protein Synthesis Initiation Inhibitor, NSC 119889, 3,4,5,6-Tetrabromofluorescein

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€275.00

€275.00


Estimated to ship on29 April 2025Details



Select a Size

Change View
25 MG
€275.00

About This Item

Empirical Formula (Hill Notation):
C20H8Br4O5
Molecular Weight:
647.89
UNSPSC Code:
12352202
NACRES:
NA.77

€275.00


Estimated to ship on29 April 2025Details


Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

light brown

solubility

DMSO: 200 mg/mL
methanol: soluble

shipped in

ambient

storage temp.

2-8°C

General description

A cell-permeable, xanthenyl compound that prevents mRNA-ribosome interaction and inhibits 5’-mediated/cap-dependent initiation of protein synthesis (>95% inhibition at 50 µM in in vitro translation assays using Krebs, rabbit reticulocyte, or E. coli S30 extract).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
human arginine methyl transferase
Product does not compete with ATP.
Reversible: no
Target IC50: 1.41 µM against human arginine methyl transferase

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Novac, O., et al. 2004. Nucleic Acids Res.32, 902.
Tan, Y.J., et al. 2003. J. Cancer Res. Clin. Oncol.129, 437.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service