Skip to Content
Merck
All Photos(1)

Key Documents

539109

Sigma-Aldrich

PAR-2 Agonist I

The PAR-2 Agonist I controls the biological activity of PAR-2. This small molecule/inhibitor is primarily used for Activators/Inducers applications.

Synonym(s):

PAR-2 Agonist I, Proteinase Activated Receptor-2 Agonist I, 2f-LIGRL-amide

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€157.00
5 MG
€318.00

€157.00


Estimated to ship on07 May 2025Details



Select a Size

Change View
1 MG
€157.00
5 MG
€318.00

About This Item

Empirical Formula (Hill Notation):
C31H53N9O7
Molecular Weight:
663.81
UNSPSC Code:
12352200
NACRES:
NA.77

€157.00


Estimated to ship on07 May 2025Details


Quality Level

Assay

≥95% (HPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

Native proteinase activated receptor-2- (PAR-2) specific agonist, SLIGRL-NH2, modified with an N-terminal furoyl group to increase its potency and resistance to metabolic degradation. Shown to activate [Ca2+]i mobilization in PAR-2-expressing cells (EC50 = 253 nM in HCT-15 cells) in vitro. Also induces PAR-2-mediated salivation (ED50 = 30 nmol/kg, i.v.) in vivo. Exhibits no effects on PAR-2-/- mice. Reactive towards human, rat, and mouse PAR-2.
The native PAR-2-specific agonist, SLIGRL-NH2, is modified with an N-terminal furoyl group to increase its potency and resistance to metabolic degradation. Shown to activate [Ca2+]i mobilization in PAR-2-expressing cells (EC50 = 253 nM using HCT-15 cells) in vitro and induce PAR-2 mediated salivation (ED50 = 30 nmol/kg, i.v.) in vivo, while exhibiting no effect on PAR-2-/- mice. Reactive towards human, rat, and murine PAR-2.

Biochem/physiol Actions

Cell permeable: no
EC50 = 253 nM as PAR-2-specific agonist using HCT-15 cells
Primary Target
PAR-2
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Sequence

2-Furoyl-Leu-Ile-Gly-Arg-Leu-NH₂

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Kawabata, A., et al. 2004. J. Pharmacol. Exp. Ther.309, 108.
Kawabata, A., et al. 2004. J. Pharmacol. Exp. Ther.309, 1098.
Ferrell, W.R., et al. 2003. J. Clin. Invest.111, 35.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service