Skip to Content
Merck
All Photos(1)

Key Documents

538927

Sigma-Aldrich

15d-PGJ2

A metabolite of PGD₂that has recently been identified as a natural ligand for PPARγ-dependent adipogenesis in transfected NIH/3T3 cells and for PPARγ response element (PPRE) in CV-1 cells.

Synonym(s):

15d-PGJ2, Prostaglandin J₂, 15-Deoxy-Δ 12,14, PPAR Agonist VII, PPARγ Agonist VI

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€230.00

€230.00


Estimated to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
1 MG
€230.00

About This Item

Empirical Formula (Hill Notation):
C20H28O3
CAS Number:
Molecular Weight:
316.43
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.77

€230.00


Estimated to ship on22 April 2025Details


Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

oil

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

solubility

DMSO: 25 mg/mL
ethanol: 25 mg/mL

shipped in

wet ice

storage temp.

−20°C

SMILES string

OC(=O)CCC\C=C/C[C@@H]1\C(=C/C=C/CCCCC)\C(=O)C=C1

InChI

1S/C20H28O3/c1-2-3-4-5-6-10-13-18-17(15-16-19(18)21)12-9-7-8-11-14-20(22)23/h6-7,9-10,13,15-17H,2-5,8,11-12,14H2,1H3,(H,22,23)/b9-7-,10-6+,18-13+/t17-/m0/s1

InChI key

VHRUMKCAEVRUBK-GODQJPCRSA-N

General description

A metabolite of PGD2 that has recently been identified as a natural ligand for PPARγ-dependent adipogenesis in transfected 3T3 cells and for PPARγ response element (PPRE) in CV-1 cells. Competes with thiazolidinediones for PPARγ binding.

Biochem/physiol Actions

Cell permeable: no
Primary Target
A natural ligand for PPARγ-dependent adipogenesis
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Harmful (C)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Petrova, T.V., et al. 1999. Proc. Natl. Acad. Sci. USA96, 4668.
Ricote, M., et al. 1998. Nature 391, 79.
Forman, B.M., et al. 1995. Cell 83, 803.
Kliewer, S.A., et al. 1995. Cell83, 813.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service