Skip to Content
Merck
All Photos(1)

Key Documents

538903

Millipore

Prostaglandin E1

Activates adenylate cyclase activity via a G-protein coupled receptor.

Synonym(s):

Prostaglandin E1, PGE₁

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€92.10

€92.10


Estimated to ship on05 May 2025Details


Request a Bulk Order

Select a Size

Change View
1 MG
€92.10

About This Item

Empirical Formula (Hill Notation):
C20H34O5
CAS Number:
Molecular Weight:
354.48
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.77

€92.10


Estimated to ship on05 May 2025Details


Request a Bulk Order

Quality Level

description

Merck USA index - 14, 7876

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white to off-white

solubility

ethanol: 50 mg/mL
methanol: 50 mg/mL
DMSO: soluble

shipped in

wet ice

storage temp.

−20°C

SMILES string

O[C@H]1[C@@H]([C@H](C(=O)C1)CCCCCCC(=O)O)\C=C\[C@@H](O)CCCCC

InChI

1S/C20H34O5/c1-2-3-6-9-15(21)12-13-17-16(18(22)14-19(17)23)10-7-4-5-8-11-20(24)25/h12-13,15-17,19,21,23H,2-11,14H2,1H3,(H,24,25)/b13-12+/t15-,16+,17+,19+/m0/s1

InChI key

GMVPRGQOIOIIMI-DWKJAMRDSA-N

General description

Activates adenylate cyclase activity via a G-protein coupled receptor.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Activates adenylate cyclase activity
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Harmful & Carcinogenic / Teratogenic (E)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for 1 month at -20°C.

Other Notes

Ammer, H. and Schulz, R. 1993. Mol. Pharmacol. 43, 556.
Rubin, D. and Laposata, M. 1991. J. Biol. Chem.266, 23618.
Kurstjens, N.P., et al. 1990. Biochem. Biophys. Res. Commun.167, 1162.
Krakauer, K.A., et al. 1986. Prostaglandins32, 301.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service