Skip to Content
Merck
All Photos(2)

Key Documents

5.38339

Sigma-Aldrich

HuR-ARE Interaction Inhibitor, CMLD-2

Synonym(s):

HuR-ARE Interaction Inhibitor, CMLD-2, 5,7-dimethoxy-8-(1-(4-methoxyphenyl)-3-oxo-3-(pyrrolidin-1-yl)propyl)-4-phenyl-2H-chromen-2-one

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€192.00

€192.00


In StockDetails



Select a Size

Change View
10 MG
€192.00

About This Item

Empirical Formula (Hill Notation):
C31H31NO6
Molecular Weight:
513.58
UNSPSC Code:
12352200
NACRES:
NA.77

€192.00


In StockDetails


Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

General description

A cell-permeable, 2H-chromen-2-one based compound that competitively binds to Hu antigen R (HuR; Ki = 350 nM) and disrupts the interaction of HuR and adenine- and uridine-rich element (ARE) of target mRNAs. Displays dose-dependent binding to full-length HuR protein and to RRM1/2 fragment. Exhibits cytotoxicity towards tumor cells and reduces their viability (IC50 = 28.9 and 18.2 µM for HCT-116 and MioPaCa2 cells, respectively). Has much reduced effect on normal WI-38 and CCD 841 CoN cells. Shown to induce cleavage of poly (ADP-ribose) polymerase and pro-caspase 3 (~ 50 µM) and reduce the half-life of Bcl-2, Msi1, and XIAP in HCT-116 cells (~20 µM) treated with actinomycin D (5 µg/ml).
A cell-permeable, 2H-chromen-2-one based compound that competitively binds to Hu antigen R (HuR; Ki = 350 nM) and disrupts the interaction of HuR and adenine- and uridine-rich element (ARE) of target mRNAs. Displays dose-dependent binding to full-length HuR protein and to RRM1/2 fragment. Exhibits cytotoxicity towards tumor cells and reduces their viability (IC50 = 28.9 and 18.2 µM for HCT-116 and MioPaCa2 cells, respectively). Has much reduced effect on normal WI-38 and CCD 841 CoN cells. Shown to induce cleavage of poly (ADP-ribose) polymerase and pro-caspase 3 (~ 50 µM) and reduce the half-life of Bcl-2, Msi1, and XIAP in HCT-116 cells (~20 µM) treated with actinomycin D (5 µg/ml).

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
HuR
Reversible: yes
Target Ki: 350 nM for HuR binding

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20&deg:). Stock solutions are stable for up to months at -20°C.

Other Notes

Wu, X., et al. 2015. ACS Chem. Biol.10, 1476.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service