Skip to Content
Merck
All Photos(1)

Key Documents

5.34335

Sigma-Aldrich

ACC1/2 Inhibitor, Cpd 9

Synonym(s):

ACC1/2 Inhibitor, Cpd 9, ACC Inhibitor, Cpd9, Acetyl-CoA Carboxylase Inhibitor, Cpd9

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€243.00

€243.00


Estimated to ship on19 May 2025Details



Select a Size

Change View
10 MG
€243.00

About This Item

Empirical Formula (Hill Notation):
C23H27N5O2
Molecular Weight:
405.49
UNSPSC Code:
12352200
NACRES:
NA.77

€243.00


Estimated to ship on19 May 2025Details


Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

tan

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

General description

A cell-permeable, spiral ketone derivative that acts as a highly potent inhibitor of acetyl-CoA carboxylase (ACC; IC50 = 27 and 33 nM for human ACC1 and ACC2, respectively). The inhibition appears to be uncompetitive with respect to ATP and non-competitive with carbonate and acetyl-CoA. Inhibits malonyl-CoA synthesis in primary rat hepatocytes (IC50 = 29.9 nM) and reduces de novo hepatic lipid synthesis (ED50 = 7.8 mg/kg in rats). Acutely reduces whole body respiratory exchange ratio (RER) to indicate a switch to fatty acid utilization. Has low protein binding in rat and dog plasma, but exhibits about 10-fold higher binding in monkey and human plasma. Displays favorable pharmacokinetic properties with good bioavailability, microsomal stability, and low plasma clearance (t1/2 = 4.8 h in rat and 5.7 h in dog, at 3 mg/kg, p.o.).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
ACC

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Griffith, D.A., et al. 2014. J. Med. Chem.57, 10512.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service