Skip to Content
Merck
All Photos(1)

Key Documents

5.33982

Sigma-Aldrich

Dock5 Inhibitor, C21

Synonym(s):

Dock5 Inhibitor, C21, N-(3,5-Dichlorophenyl)benzenesulfonamide

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
€153.00

€153.00


Estimated to ship on28 May 2025Details



Select a Size

Change View
50 MG
€153.00

About This Item

Empirical Formula (Hill Notation):
C12H9Cl2NO2S
CAS Number:
Molecular Weight:
302.18
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€153.00


Estimated to ship on28 May 2025Details


Assay

≥98% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

gray

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(Nc2cc(cc(c2)Cl)Cl)c1ccccc1

InChI

1S/C12H9Cl2NO2S/c13-9-6-10(14)8-11(7-9)15-18(16,17)12-4-2-1-3-5-12/h1-8,15H

InChI key

HMKZVAZQKOKXRZ-UHFFFAOYSA-N

General description

A cell-permeable, bioavailable, non-toxic benzenesulfonamide compound that directly blocks Dock5-mediated Rac activation (kcat/KM = 7.9 x 104 & 9.5 x 104 M-1s-1 for Rac1 & Rac2 by Dock5-DHR2, respectively) with selectivity over RhoA and Cdc42. At higher concentrations, affects Dock1 and Dock2 guanine nucleotide exchange factor activity. Shown to reversibly perturb podosome organization and inhibit osteoclastic bone degradation in a dose-dependent manner. Efficiently prevents both arthritis and metastasis-induced bone loss while preserving bone formation in several osteolytic disease mouse model (25 mg/kg, i.p. or i.v., 5 days a week for 4 weeks). Unlike alendronate (Cat. No. 126855), C21 does not impair bone formation.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Dock5

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Vives, V., et al. 2015. Nat. Commun.6, 6218.
Vives, V., et al. 2011. J. Bone Miner. Res.26, 1099.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service