Skip to Content
Merck
All Photos(1)

Key Documents

5.32757

Sigma-Aldrich

RIPK2 Inhibitor, OD36

Synonym(s):

RIPK2 Inhibitor, OD36, RIPK2 inhibitor, RIP kinase inhibitor

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€131.00

€131.00


Estimated to ship on25 February 2025Details


Request a Bulk Order

Select a Size

Change View
5 MG
€131.00

About This Item

Empirical Formula (Hill Notation):
C16H15ClN4O2
CAS Number:
Molecular Weight:
330.77
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€131.00


Estimated to ship on25 February 2025Details


Request a Bulk Order

Assay

≥97% (HPLC)

Quality Level

form

powder

potency

5.3 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

light beige

solubility

DMSO: 50 mg/mL

storage temp.

−20°C

General description

A cell permeable, highly potent, selective, and ATP-competitive inhibitor of receptor-interacting protein 2 (RIPK2, IC₅₀ = 5.3 nM).
A cell permeable, pyrazolopyridine containing macrocycle derivative that acts as a highly potent, selective, ATP-competitive and reversible inhibitor of receptor-interacting protein 2 (RIPK2, IC50 = 5.3 nM). Shown to minimally affect the activity of 366 other protein kinases. At higher concentrations (100 nM), inhibits ALK-2, salt-inducible kinase 2 (SIK2), activin receptor type-2B (ACVR2B), and activin A receptor type II-like 1 (ACVRL1) by ~80%. Blocks RIPK2 autophosphorylation and downstream NF-κB and MAP kinase signaling induced by muramyl dipeptide (MDP), and reduces MDP-induced cellular infiltration of neutrophils and lymphocytes in an in vivo peritonitis model (6.25 mg/kg, i.p.).

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.
A cell permeable, pyrazolopyrinide containing macrocycle derivative that acts as a highly potent, selective, and ATP-competitive inhibitor of receptor-interacting protein 2 (RIPK2, IC50 = 5.3 nM). At ~20-fold higher concentration inhibits (~80% inhibition at 100 nM) the activity of ALK-2, salt-inducible kinase 2 (SIK2), activin receptor type-2B (ACVR2B), and activin A receptor type II-like 1(ACVRL1), but does not affect the activity of 366 other protein kinases. Blocks RIPK2 autophosphorylation and downstream NF-κB and MAP kinase signaling induced by muramyl dipeptide (MDP) and reduces MDP-induced cellular infiltration of neutrophils and lymphocytes in an in vivo peritonitis model (6.25 mg/kg).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
RIPK2

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Tigno-Aranjuez, J.T., et al. 2014. J. Biol. Chem.289, 29651.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service