Skip to Content
Merck
All Photos(1)

Key Documents

5.31711

Sigma-Aldrich

DOT1L Inhibitor, SYC-522

Synonym(s):

DOT1L Inhibitor, SYC-522, Histone methyltransferase DOT1L Inhinitor, SYC-522

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€207.00

€207.00


In StockDetails


Request a Bulk Order

Select a Size

Change View
5 MG
€207.00

About This Item

Empirical Formula (Hill Notation):
C27H40N8O4
CAS Number:
Molecular Weight:
540.66
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€207.00


In StockDetails


Request a Bulk Order

Assay

≥98% (HPLC)

Quality Level

form

solid

potency

500 pM Ki

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white to light yellow

solubility

DMSO: 50 mg/mL

storage temp.

−20°C

General description

A cell permeable S-adenosyl-L-methionine (SAM) derivative that acts as a highly potent and selective inhibitor of histone 3-lysine79 (H3K79) methyltransferase DOT1L (Ki = 500 pM ) and inhibits H3K79 methylation. Does not affect the activity of PRMT1, CARM1 and SUV39H1 (IC50 >100 µM). Blocks cell cycle at the G0/G1 phase. Although it does not induce apoptosis, it sensitizes MLL rearranged leukemia cells to chemotherapeutic agents (mitoxantrone, etoposide, cytarabine) to cause apoptotic cell death. Shown to down-regulate the expression of HOXA9 and MEIS1, leukemia-relevant genes, by over 50%.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
DOT1L

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Liu, W., et al. 2014. PLoS One.9, e98270.
Anglin, J.L., et al. 2012. J. Med. Chem.55, 8066.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service