Skip to Content
Merck
All Photos(1)

Documents

5.31371

Sigma-Aldrich

S1P Receptor 2 Agonist, CYM-5520

Synonym(s):

S1P Receptor 2 Agonist, CYM-5520, S1PR2 Agonist, CYM-5520, Sphingosine-1-Phosphate Receptor 2 Agonist, CYM 5520, EDG5 Agonist

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H19N3O2
CAS Number:
Molecular Weight:
345.39
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

General description

A pyrrolyl ketone derivative that acts as a potent, selective, allosteric agonist of sphingosine-1-phosphate receptor 2 (S1PR2; EC50 = 480 nM) that does not replace native ligand and its binding is not competitive with JTE-013. Does not affect the activity of S1PR1, 3, and 5 and the activity of 29 other receptors and transports in any significant manner. Acts as a full agonist for both wild type and triple mutant S1PR2 (EC50 = 1.6 and 1.5 µM, respectively).
A pyrrolyl ketone derivative that acts as a potent, selective, allosteric agonist of sphingosine-1-phosphate receptor 2 (S1PR2; EC50 = 480 nM) that does not replace native ligand and its binding is not competitive with JTE-013. Does not affect the activity of S1PR1, 3, and 5 and the activity of 29 other receptors and transports in any significant manner. Acts as a full agonist for both wild type and triple mutant S1PR2 (EC50 = 1.6 and 1.5 µM, respectively).

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Primary Target
S1PR2
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C).

Other Notes

Satsu, H., et al. 2013. Bioorg. Med. Chem.21, 5373.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hideo Satsu et al.
Bioorganic & medicinal chemistry, 21(17), 5373-5382 (2013-07-16)
Molecular probe tool compounds for the Sphingosine 1-phosphate receptor 2 (S1PR2) are important for investigating the multiple biological processes in which the S1PR2 receptor has been implicated. Amongst these are NF-κB-mediated tumor cell survival and fibroblast chemotaxis to fibronectin. Here

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service