Skip to Content
Merck
All Photos(1)

Key Documents

5.30748

Sigma-Aldrich

SRT1720 HCl

InSolution, ≥98%, 25 mM in DMSO

Synonym(s):

InSolution SRT1720, HCl, N-(2-(3-(1-Piperazinylmethyl)imidazo[2,1-b]thiazol-6-yl)phenyl)-2-quinolinecarboxamide, 4HCl;, Histone Acetyltransferase Inhibitor XI, p300/CBP Inhibitor IX, SRT1720, HCl InSolution, HAT Inhibitor XI, SIRT3 Inhibitor II, Sirtuin-3 In

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€243.00

€243.00


Estimated to ship on07 April 2025Details


Request a Bulk Order

Select a Size

Change View
10 MG
€243.00

About This Item

Empirical Formula (Hill Notation):
C25H23N7OS · xHCl
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€243.00


Estimated to ship on07 April 2025Details


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles
desiccated (hygroscopic)
protect from light

storage temp.

−70°C

SMILES string

[s]1c2[n](c(c1)CN6CCNCC6)cc(n2)c3c(cccc3)NC(=O)c4nc5c(nc4)cccc5.Cl

InChI

1S/C25H23N7OS.ClH/c33-24(22-13-27-20-7-3-4-8-21(20)28-22)29-19-6-2-1-5-18(19)23-15-32-17(16-34-25(32)30-23)14-31-11-9-26-10-12-31;/h1-8,13,15-16,26H,9-12,14H2,(H,29,33);1H

InChI key

DTGRRMPPXCRRIM-UHFFFAOYSA-N

General description

An enhanced aqueous soluble version of SRT1720 (Cat. No. 567860 ).

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
SIRT3
Reversible: yes
Secondary Target
p300 HAT

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Physical form

A 25 mM (10 mg/650 µl) sterile-filtered solution of SRT1720, HCl (Cat. No. 567860 ) in DMSO.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C). Aliquots are stable for up to 6 months at -20°C.

Other Notes

Baur, J.A., et al. 2012. Nat. Rev. Drug Discov.11, 443.
Minor, R.K., et al. 2011. Sci. Rep.1, 70.

Huber, J.L., et al. 2010. Future Med. Chem.2, 1751.
Pacholec, M., et al. 2010. J. Biol. Chem.285, 8340.
Jin, L., et al. 2009. Protein Sci.18, 514.
Feige, J.N., et al. 2008. Cell Metab.8, 347.
Milne, J.C., et al. 2007. Nature450, 712.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

188.6 °F

Flash Point(C)

87 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service