Skip to Content
Merck
All Photos(1)

Key Documents

5.30538

Sigma-Aldrich

cPLA2α Inhibitor II, Pyrrophenone

Synonym(s):

cPLA2α Inhibitor II, Pyrrophenone, Phospholipase A2α Inhibitor

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€227.00

€227.00


Estimated to ship on20 May 2025Details


Request a Bulk Order

Select a Size

Change View
1 MG
€227.00

About This Item

Empirical Formula (Hill Notation):
C49H37F2N3O5S2
CAS Number:
Molecular Weight:
849.96
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€227.00


Estimated to ship on20 May 2025Details


Request a Bulk Order

Assay

≥98% (HPLC)

Quality Level

form

powder

potency

4.2 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 100 mg/mL

storage temp.

−20°C

General description

Pyrrophenone is a cell-permeable, non-toxic, pyrolidine derivative that acts as a highly potent, and reversible inhibitor of cytosolic phospholipase A2a (cPLA2a; IC50 = 4.2 nM). Does not inhibit the activities of PLA IB and IIA groups even at higher concentrations (~250 µM). Unlike AACOCF3 (Cat. No. 100109), its action is shown to be rapid and does not require a pre-incubation period. Diminishes the release of arachidonic acid (IC50 = 24 nM), prostaglandin E2 (PGE2; IC50 = 25 nM) and leukotriene C4 (IC50 = 14 nM) in A23187 stimulated THP-1 cells. Its effects on production of PGE2 and thromboxane B2 are comparable to those of indomethacin. Also shown to inhibit the lysophospholipase and esterase activity of cPLA2&alpha.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
PLA2a

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service