Skip to Content
Merck
All Photos(1)

Key Documents

530070

Sigma-Aldrich

Pristimerin

A cell-permeable plant dienone-phenolic triterpenoid, naturally isolated from tripterygium wilfordii.

Synonym(s):

Pristimerin, Monoacylglycerol Lipase Inhibitor II, Proteasome Inhibitor XV, NF-κB Activation Inhibitor IX, MAGL Inhibitor II

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€98.60

€98.60


Estimated to ship on17 February 2025Details



Select a Size

Change View
5 MG
€98.60

About This Item

Empirical Formula (Hill Notation):
C30H40O4
CAS Number:
Molecular Weight:
464.64
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€98.60


Estimated to ship on17 February 2025Details


Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

red

solubility

DMSO: 10 mg/mL
ethanol: 2.5 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C30H40O4/c1-18-19-8-9-22-28(4,20(19)16-21(31)24(18)32)13-15-30(6)23-17-27(3,25(33)34-7)11-10-26(23,2)12-14-29(22,30)5/h8-9,16,23,32H,10-15,17H2,1-7H3/t23-,26-,27-,28+,29-,30+/m1/s1

InChI key

JFACETXYABVHFD-WXPPGMDDSA-N

General description

A cell-permeable plant dienone-phenolic triterpenoid, naturally isolated from tripterygium wilfordii, whose inhibitory activities against proteasome chymotrypsin subunit (IC50 = 2.2 and 3.0 µM against purified rabbit 20S and human 26S proteasome, respectively) and cellular NF-κB pathway/IKK activation most likely account for its reported anticancer efficacy both in vitro (IC50 ≤610 nM in A549, K562, KBM5, MCF-7, MDA-MB-231, HepG2, and primary human leukemia cell cultures) and in mice in vivo (1 to 2.5 mg/kg/day, s.c.), including imatinib/STI571-resistant cancer cells harboring Bcr-Abl T315I mutation. Also reported to inhibit the activity of monoacylglycerol lipase (MGL), the enzyme that hydrolyzes and deactivates the cannabionoid receptor ligand 2-Arachidonoyl-sn-glycerol (2-AG), in a reversible manner with high potency (IC50 = 93 nM).

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Lu, Z., et al. 2010. Mol. Cancer9, 112.
King, A.R., et al. 2009. Chem. Biol.16, 1045.
Tiedemann, R.E., et al. 2009. Blood113, 4027.
Yang, H., et al. 2008. J. Cell. Biochem.103, 234.
Wu, C.C., et al. 2005. Mol. Cancer Ther.4, 1277.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service