Skip to Content
Merck
All Photos(1)

Key Documents

530006

Sigma-Aldrich

Prion Protein, His•Tag®, Human, Recombinant, E. coli

Synonym(s):

rhPrP c, rhPrPc

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€908.00

€908.00


Estimated to ship on17 April 2025Details



Select a Size

Change View
100 μG
€908.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

€908.00


Estimated to ship on17 April 2025Details


Assay

≥95% (SDS-PAGE)

Quality Level

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

wet ice

storage temp.

−20°C

General description

Recombinant, human prion protein (amino acids 23-231) fused to a His•Tag sequence and expressed in E. coli. Prion diseases or transmissible spongiform encephalopathies are neurodegenerative diseases that affect both humans and animals. All prion diseases share the same molecular pathogenic mechanism that involves conversion of normal cellular prion protein (PrPc) into a form that is insoluble in non-ionic detergent and partially resistant to proteases (PrPsc). Useful as an antigen standard in immunochemical detection of Creutzfeldt-Jakob Disease (CJD).

Warning

Toxicity: Standard Handling (A)

Physical form

In 10 mM sodium acetate, pH 4.0.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

Calzolai, L., and Zahn, R. 2003. J. Biol. Chem.278, 35592.
Prusiner, S.B. 1998. Proc. Natl. Acad. Sci. USA95, 13363.
Swietnicki, W., et al. 1997. J. Biol. Chem.272, 27517.
Pan, K.M., et al. 1993. Proc. Natl. Acad. Sci. USA90, 10962.
Basler, K., et al. 1986. Cell46, 417.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
HIS TAG is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service