Skip to Content
Merck
All Photos(2)

Key Documents

529659

Sigma-Aldrich

DDD00033325

≥95% (HPLC), syrup, single hESC survival promoter, Calbiochem®

Synonym(s):

Pro-survival Compound, DDD00033325, DDD00033325

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€230.00

€230.00


In StockDetails


Request a Bulk Order

Select a Size

Change View
10 MG
€230.00

About This Item

Empirical Formula (Hill Notation):
C17H19N5
CAS Number:
Molecular Weight:
293.37
UNSPSC Code:
51111800
NACRES:
NA.77

€230.00


In StockDetails


Request a Bulk Order

Product Name

Pro-survival Compound, DDD00033325, A cell-permeable, pyrrolidine small molecule that promotes single human embryonic stem cell survival up to 20-fold over background with equal or higher potency than Y-27632.

Quality Level

Assay

≥95% (HPLC)

form

syrup

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 100 mg/mL

shipped in

wet ice

storage temp.

−20°C

SMILES string

C12=CC=NN1C=CC(N[C@@H]3CCN(CC4=CC=CC=C4)C3)=N2

InChI

1S/C17H19N5/c1-2-4-14(5-3-1)12-21-10-7-15(13-21)19-16-8-11-22-17(20-16)6-9-18-22/h1-6,8-9,11,15H,7,10,12-13H2,(H,19,20)/t15-/m1/s1

InChI key

ZJVYTXWKNDTQNC-OAHLLOKOSA-N

General description

A cell-permeable, pyrrolidine small molecule that promotes single human embryonic stem cell survival up to 20-fold over background (EC50 = 3.9 M), with equal or higher potency than Y-27632 (Cat. No. 688000) in 5 cell lines (SA461, SA181, SA167, SA121 and SA002) tested at 10 M. An in vitro high-content screening assay, which profiles a panel of more than 70 protein kinases, indicates that this small molecule is more selective than Y-27632 and only displays an inhibitory effect toward ROCK2 (>83% inhibition, ROCK1 was not included in the panel) and PRK2 (76% inhibition) at 10 M. Furthermore, it exhibits less cytotoxicity at a greater concentration of 30 M than Y-27632 without marked adverse effects for up to 120 hours.

Packaging

Packaged under inert gas

Warning

Toxicity: Regulatory Review (Z)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C. Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Andrews, P.D., et al. 2010. Biochem. J.432, 21.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service