Skip to Content
Merck
All Photos(1)

Key Documents

528281

Sigma-Aldrich

Polo-like Kinase Inhibitor IV, SBE13

The Polo-like Kinase Inhibitor IV, SBE13, also referenced under CAS 775294-82-1, controls the biological activity of Polo-like Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

Polo-like Kinase Inhibitor IV, SBE13, N-(4-((6-Chloro-3-pyridinyl)methoxy)-3-methoxybenzyl)-N-(2-(3,4-dimethoxyphenyl)ethyl)amine

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€122.00

€122.00


Estimated to ship on22 April 2025Details



Select a Size

Change View
10 MG
€122.00

About This Item

Empirical Formula (Hill Notation):
C24H27ClN2O4
CAS Number:
Molecular Weight:
442.94
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€122.00


Estimated to ship on22 April 2025Details


Quality Level

Assay

≥94% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

beige

solubility

DMSO: 25 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A cell-permeable vanillin-derived pyridyl-ethanamine compound that acts as a highly potent type II Plk1 inhibitor (IC50 of 200 pM) and targets the less conserved DFG motif that is only accessible when the kinase is in its inactive DFG-out conformation, resulting in an excellent selectivity over Plk2 (IC50 >66 µM), Plk3 (IC50 = 875 nM), and aurora A kinase (no detectable effect). Shown to inhibit the proliferation of HeLa cells with an EC50 of 18 µM.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Keppner, S., et al. 2009. ChemMedChem.4, 1806.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service