Skip to Content
Merck
All Photos(1)

Documents

528180

Sigma-Aldrich

Plasminogen, Glu-Type, Human Plasma

Synonym(s):

Glu-type plasminogen, Plasminogen

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.25

form

liquid

Quality Level

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

foreign activity

Plasmin, none detected

shipped in

wet ice

storage temp.

−70°C

General description

Native Glu-type plasminogen from human plasma. Plasminogen is a single chain glycoprotein found in normal plasma at ~12 mg per deciliter, and is the inactive precursor of the serine protease plasmin.
Native Glu-type plasminogen from human plasma. Plasminogen is a single chain glycoprotein zymogen, which is synthesized in the liver and circulates in plasma at a concentration of approximately 2.4 µM. The plasminogen molecule contains 790 amino acids, 24 disulfide bridges, no free sulfhydryls, and 5 regions of internal sequence homology, known as kringles, between Lys77 and Arg560. These kringle regions are homologous to the kringle domains in t-PA, u-PA, and prothrombin. Native Glu-plasminogen is readily converted to Lys77-plasminogen by plasmin hydrolysis of the Lys76-Lys77 peptide bond. The plasminogen is converted to plasmin by cleavage at Arg540-Val561 by urokinase, streptokinase, or tissue plasminogen activator.

Warning

Toxicity: Irritant (B)

Physical form

In 100 mM NaCl, 50 mM Tris-HCl, pH 7.5.

Preparation Note

Prepared from plasma that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Analysis Note

Single band by SDS-PAGE

Other Notes

Collen, D. 1986. In Blood Coagulation (Zwaal, R.E.A., and Hemker, H.C. eds.) Elsevier, N.Y. p. 243.
Barlow, G.H., et al. 1984. Biochemistry23, 2384.
Wohl, R.C., et al. 1982. Thromb. Res.27, 523.
Castellino, F.J., et al. 1981. Methods Enzymol.80, 365.
Robbins, K.C. 1976. Methods Enzymol.45, 257.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service