Skip to Content
Merck
All Photos(1)

Key Documents

528118

Sigma-Aldrich

PI 3-Kδ Inhibitor X, IC87114

The PI 3-Kδ Inhibitor X, IC87114, also referenced under CAS 371242-69-2, controls the biological activity of PI 3-Kδ. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

PI 3-Kδ Inhibitor X, IC87114, Phosphatidylinositol 3-Kinase p110d Inhibitor, PI 3-K Inhibitor X

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€162.00

€162.00


Estimated to ship on23 April 2025Details



Select a Size

Change View
5 MG
€162.00

About This Item

Empirical Formula (Hill Notation):
C22H19N7O
CAS Number:
Molecular Weight:
397.43
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€162.00


Estimated to ship on23 April 2025Details


Quality Level

Assay

≥95% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

DMSO: 5 mg/mL

shipped in

wet ice

storage temp.

−20°C

SMILES string

[n]4(c5ncnc(c5nc4)N)Cc1[n]([c](c3c(n1)cccc3C)=O)c2c(cccc2)C

InChI

1S/C22H19N7O/c1-13-6-3-4-9-16(13)29-17(27-15-8-5-7-14(2)18(15)22(29)30)10-28-12-26-19-20(23)24-11-25-21(19)28/h3-9,11-12H,10H2,1-2H3,(H2,23,24,25)

InChI key

GNWHRHGTIBRNSM-UHFFFAOYSA-N

General description

A cell permeable, selective inhibitor of PI 3-Kδ (IC50= 1.82, 0.07, and 1.24 for p110b, p110d, and p110g, respectively, but may also target p110a at concentrations as high as 100 µM with less potency) in NB-4 and primary APL cells. It has been reported that the constitutive activation of PI3K/Akt signaling in primary AML cells is predominantly attributable to PI 3-Kδ. Pharmacologic use of PI 3-Kδ inhibitor has been shown to block PI3K/Akt/mTOR signaling and could thereby reduce proliferation and induce apoptosis. A related study further suggests PI 3-Kδ inhibitor as a key therapeutic target among PI 3-K isoforms for allergy- and mast cell-related diseases.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Billottet, C., et al. 2009. Cancer Res.69,1027.
Ali, K., et al. 2008. J. Immunol.180, 2538.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yukai Jing et al.
Science advances, 6(17), eaax9455-eaax9455 (2020-06-05)
The adaptor protein, STING (stimulator of interferon genes), has been rarely studied in adaptive immunity. We used Sting KO mice and a patient's mutated STING cells to study the effect of STING deficiency on B cell development, differentiation, and BCR

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service