Skip to Content
Merck
All Photos(1)

Key Documents

528111

Sigma-Aldrich

PI 3-Kα Inhibitor IV

The PI 3-Kα Inhibitor IV, also referenced under CAS 1188890-32-5, controls the biological activity of PI 3-Kα. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

PI 3-Kα Inhibitor IV, 3-(4-Morpholinothieno[3,2-d]pyrimidin-2-yl)phenol, 2HCl, PI 3-K Inhibitor IV

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€286.00

€286.00


Estimated to ship on01 April 2025Details



Select a Size

Change View
5 MG
€286.00

About This Item

Empirical Formula (Hill Notation):
C16H15N3O2S · 2HCl
CAS Number:
Molecular Weight:
386.30
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€286.00


Estimated to ship on01 April 2025Details


Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

off-white

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[s]1c2c(nc(nc2N4CCOCC4)c3cc(ccc3)O)cc1.Cl.Cl

InChI key

WPSXNMUJNQUDND-UHFFFAOYSA-N

General description

A cell-permeable morpholino-thienopyrimidine compound that acts as a potent and isoform-selective inhibitor of PI 3-kinases (IC50 = 2 nM, 16 nM, 660 nM, and 220 nM for p110α, p110β p110γ, and PI 3-K C2β, respectively) and inhibits non-PI 3-K kinases only at much higher concentrations (IC50 ≥3.4 µM for Cdk2/E, KDR, PKA, and PKCα). Shown to inhibit cell proliferation (IC50 = 580 nM) and serum-stimulated Akt phosphorylation (IC50<3 µM) in A375 melanoma cells.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
PI 3-Kα
Product does not compete with ATP.
Reversible: no
Target IC50: 2 nM, 16 nM, 660 nM, and 220 nM for p110α, p110β p110γ, and PI 3-K C2β, respectively

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Hayakawa, M., et al. 2006. Bioorg. Med. Chem.14, 6847.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service