Skip to Content
Merck
All Photos(1)

Key Documents

524751-M

Sigma-Aldrich

Phosphodiesterase 7A1, Human, Recombinant, S. frugiperda

Synonym(s):

hPDE7A1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Pricing and availability is not currently available.

recombinant

expressed in baculovirus infected Sf21 cells

Quality Level

form

liquid

specific activity

≥0.5 U/μL

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

storage temp.

−70°C

General description

Full-length, recombinant, human phosphodiesterdase 7A1 (PDE7A1) expressed in S. frugiperda insect cells using a baculovirus expression system. PDE7A1 is required for T-cell activation induced by the costimulation of CD3 and CD28 receptors. Increased PDE7A1 correlates with a decrease in cAMP, increased IL-2, and increased cell proliferation. Supplied as a partially purified preparation of catalytically active enzyme useful for inhibitor and activator assays.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of enzyme that is required to convert 1 pmol cAMP to AMP per min at 30°C, pH 7.5.

Physical form

In 150 mM NaCl, 10 mM Tris, 0.1% NP-40, pH 8.3.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Han, P., et al. 2006. J. Biol. Chem.281, 15050.
Lugnier, C. 2006. Pharmacol Ther.109, 366.
Bloom, T.J., and Beavo, J.A. 1996. Proc. Natl. Acad. Sci. U S A93, 14188.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

EU REACH Annex XIV (Authorisation List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service