524736
Phosphodiesterase 4B2, His•Tag®, Human, Recombinant, S. frugiperda
Synonym(s):
PDE4B2, cAMP-Specific Phosphodiesterase 4B2
Select a Size
€465.00
Estimated to ship on16 April 2025Details
Select a Size
About This Item
€465.00
Estimated to ship on16 April 2025Details
Recommended Products
recombinant
expressed in baculovirus infected Spodoptera frugiperda cells
Quality Level
form
liquid
specific activity
≥0.25 U/μL
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
avoid repeated freeze/thaw cycles
shipped in
wet ice
storage temp.
−70°C
General description
Warning
Unit Definition
Physical form
Reconstitution
Other Notes
Smith, P.G., et al. 2005 Blood105, 308.
Baroja, M.L., et al. 1999. J. Immunol.162, 2016.
Legal Information
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
EU REACH SVHC Candidate List
EU REACH Annex XVII (Restriction List)
EU REACH Annex XIV (Authorisation List)
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service