Skip to Content
Merck
All Photos(1)

Key Documents

524650

Millipore

PBS Tablets

10 mM phosphate buffer, and 3 mM KCl, tablet, pH 7.4 at 25°C

Synonym(s):

PBS, Phosphate Buffered Saline Tablets

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€66.20

€66.20


Estimated to ship on04 April 2025Details


Request a Bulk Order

Select a Size

Change View
1 EA
€66.20

About This Item

UNSPSC Code:
12161706
NACRES:
NA.25

€66.20


Estimated to ship on04 April 2025Details


Request a Bulk Order

Product Name

PBS Tablets, Dissolving one tablet of PBS buffer in 1 liter of deionized H2O yields 140 mM NaCl, 10 mM phosphate buffer, and 3 mM KCl, pH 7.4 at 25°C. Note: 1 each = 10 tablets.

Quality Level

form

tablet

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

shipped in

ambient

storage temp.

10-30°C

General description

Developed for use in immunoassay applications. Dissolving one tablet in 1 liter of deionized H2O yields 140 mM NaCl, 10 mM phosphate buffer, and 3 mM KCl, pH 7.4 at 25°C. Note: 1 each = 10 tablets.
Dissolving one tablet in 1 liter of deionized H2O yields 140 mM NaCl, 10 mM phosphate buffer, and 3 mM KCl, pH 7.4 at 25°C. Note: 1 each = 10 tablets.

Warning

Toxicity: Irritant (B)

Preparation Note

Dissolve one tablet in one liter dH₂O to yield 10 mM phosphate buffer, pH 7.4, 140 mM NaCl, 3 mM KCl.

Reconstitution

Following reconstitution, store in the refrigerator (4°C) or filter sterilize and store at room temperature (20°C).

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Valentina Agnolon et al.
Scientific reports, 10(1), 3016-3016 (2020-02-23)
Circulating HER2 extracellular domain (HER2 ECD) levels were proposed as a surrogate for HER2 tissue expression to monitor breast cancer patients for early relapse or responses to standard or HER2-targeted therapies, such as the monoclonal antibody (mAb) trastuzumab. Currently, available commercial

Articles

Antibody fragmentation with our pepsin digestion protocol for IgG antibody fragmentation and preparation of F(ab’).

Questions

1–4 of 4 Questions  
  1. Is this product sterile/ Is it usable in the context of microbiological analysis ?

    1 answer
    1. Product 524650 is not guaranteed to be sterile and has not been tested for use with cell culture. Sterile filtered solution forms of PBS are available that have been assayed for suitability with cell culture. Please see products P2272, P4474, 806544, and 6506-OP.

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  4. Does your product code 524650 contain calcium or magnesium?

    1 answer
    1. The formulation of this product is considered proprietary. However, this item does not contain calcium or magnesium.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service