Skip to Content
Merck
All Photos(1)

Key Documents

52125BC

Millipore

FLASH® 28 Tube Rack for Digital Dry Bath Heater

BioControl, For use with FLASH® Digital Dry Bath Heater

Sign Into View Organizational & Contract Pricing

Select a Size

1 UNIT
€125.00

€125.00


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
1 UNIT
€125.00

About This Item

UNSPSC Code:
41104424

€125.00


Please contact Customer Service for Availability

Request more information

manufacturer/tradename

BioControl Systems 52125
FLASH®

storage condition

room temperature

application(s)

food and beverages

compatibility

for use with FLASH (63003/63003BC)

storage temp.

room temp

General description

Holds 28 x 10 mm tubes

Components

One block

Compatibility

Compatible with Digital Dry Bath Heater, 73011BC, 73061BC

Testing Method

PCR

Legal Information

FLASH is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service