Skip to Content
Merck
All Photos(1)

Key Documents

5.09890

Sigma-Aldrich

Benztropine

Synonym(s):

Benztropine, Benzatropine, Cogentin

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
€78.00

€78.00


Estimated to ship on22 May 2025Details


Request a Bulk Order

Select a Size

Change View
50 MG
€78.00

About This Item

Empirical Formula (Hill Notation):
C21H2NO · xCH4O3S
CAS Number:
Molecular Weight:
284.25 (free base basis)
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€78.00


Estimated to ship on22 May 2025Details


Request a Bulk Order

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(O)C.N1([C@@H]2CC[C@@H]1CC(C2)OC(c4ccccc4)c3ccccc3)C

InChI

1S/C21H25NO.CH4O3S/c1-22-18-12-13-19(22)15-20(14-18)23-21(16-8-4-2-5-9-16)17-10-6-3-7-11-17;1-5(2,3)4/h2-11,18-21H,12-15H2,1H3;1H3,(H,2,3,4)/t18-,19-;/m1./s1

InChI key

CPFJLLXFNPCTDW-STYNFMPRSA-N

General description

A brain penetrating azabicyclo[3.2.1]octane derivative with anti-cholinergic and anti-histaminergic properties. Acts as an antagonist of muscarinic M1 and M3 receptors induces robust differentiation of oligodendrocytes precursor cell differentiation (OPC; EC50 = 500 nM) and promotes myelination. Reduces the expression of Notch 1 in immature OPC and reduces the levels of cyclin D1 and D2 and c-Fos and c-Jun indicating its inhibitory effect on cell cycle progression. Shown to reduce the severity of the acute phase of multiple sclerosis and virtually eliminates the relapse phase in animal models. Significantly reduces the severity in the experimental autoimmune encephalomyelitis (EAE) model of relapsing-remitting multiple sclerosis when administered alone or co-administered with FTY720 (Cat. No. 344597).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Muscarinic M1/M3
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Toxic (F)

Other Notes

Deshmukh, V.A, et al. 2013. Nature 502,327.
Doshay, L.J., et al. 1956. J Am Med Assoc.162, 1031.
Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service