Skip to Content
Merck
All Photos(1)

Documents

5.08436

Sigma-Aldrich

AEBSF Hydrochloride

InSolution, ≥97%, Specific, irreversible inhibitor of serine proteases

Synonym(s):

InSolution AEBSF, Hydrochloride, 4-(2-Aminoethyl)benzenesulfonylfluoride, HCl, 4-(2-Aminoethyl)benzene-1-sulfonylfluoride, HCl, AEBSF, HCl InSolution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H10FNO2S · xHCl
CAS Number:
Molecular Weight:
203.23 (free base basis)
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles
protect from light

storage temp.

−70°C

InChI

1S/C8H10FNO2S.ClH/c9-13(11,12)8-3-1-7(2-4-8)5-6-10;/h1-4H,5-6,10H2;1H

InChI key

WRDABNWSWOHGMS-UHFFFAOYSA-N

General description

Specific, irreversible inhibitor of serine proteases. Inhibits chymotrypsin, kallikrein, plasmin, thrombin, trypsin, and related thrombolytic enzymes. Stable, nontoxic alternative to PMSF (Cat. No. 52332) and DFP (Cat. No. 30967). A β-secretase inhibitor that inhibits β-amyloid peptide (Aβ) production and enhances amyloid precursor protein (sAPPUα) secretion in several cell lines at millimolar concentrations. Use at the same molar concentrations (0.1-1.0 mM) as PMSF for most applications.

Biochem/physiol Actions

Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Physical form

A 50 mM (50 mg/4.17 ml) sterile-filtered solution of AEBSF, Hydrochloride (Cat. No. 101500) in H₂O.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C). Aliquots are stable for up to 1 year at -20°C.

Other Notes

Citron, M., et al. 1996. Neuron17, 171.
Ohkubo, I., et al. 1994. J. Biochem.116, 1182.
Mintz, G.R. 1993. BioPharm.6, 34.
Lawson, W.B., et al. 1982. Folia Haematol.109, 52.
Markwardt, F., et al. 1974. Biochem. Pharmacol.23, 2247.
Markwardt, F., et al. 1973. Thromb. Res.2, 343.

Baker, B.R., and Cory, M. 1971. J. Med. Chem.14, 119.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service