Skip to Content
Merck
All Photos(1)

Key Documents

5.08318

Sigma-Aldrich

SHIP2 Inhibitor II, CPDA

Synonym(s):

SHIP2 Inhibitor II, CPDA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H15ClF2N2O2
CAS Number:
Molecular Weight:
388.80
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

General description

A cell-permeable aminopyridine acetamide based compound that acts as an inhibitor of SH2 domain-containing inositol 5′-phosphatase 2 (SHIP2) that can overcome insulin resistance in 3T3-L1 adipocytes. Enhances insulin-stimulated Akt phosphorylation (Thr308) in TNFa-treated 3T3-L1 adipocytes (~10 µM) and in primary cultured neurons from rat cerebral cortex. Improves glucose tolerance in db/db mice (~300 mg/kg b.i.d) without affecting their fasting glucose levels. Also reduces the expression levels of glucose-6-phosphatase and phosphoenolpyruvate carboxykinase.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
SHIP2

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Ichihara, Y., et al, 2013. Eur. J. Med. Chem.62, 649.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service