Skip to Content
Merck
All Photos(1)

Key Documents

506121

Sigma-Aldrich

p38 MAP Kinase Inhibitor III

The p38 MAP Kinase Inhibitor III, also referenced under CAS 581098-48-8, controls the biological activity of p38 MAP Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

p38 MAP Kinase Inhibitor III, ML3403, (RS)-{4-[5-(4-Fluorophenyl)-2-methylsulfanyl-3H-imidazol-4-yl]pyridin-2-yl}-(1-phenylethyl)amine]

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€223.00

€223.00


Estimated to ship on24 April 2025Details



Select a Size

Change View
1 MG
€223.00

About This Item

Empirical Formula (Hill Notation):
C23H21FN4S
CAS Number:
Molecular Weight:
404.50
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.54

€223.00


Estimated to ship on24 April 2025Details


Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

Fc1ccc(cc1)c2[nH]c(nc2c3cc(ncc3)NC(C)c4ccccc4)SC

InChI

1S/C23H21FN4S/c1-15(16-6-4-3-5-7-16)26-20-14-18(12-13-25-20)22-21(27-23(28-22)29-2)17-8-10-19(24)11-9-17/h3-15H,1-2H3,(H,25,26)(H,27,28)

InChI key

VXPWQNBKEIVYIS-UHFFFAOYSA-N

General description

A cell-permeable methylsulfanylimidazole compound that acts as a potent, selective, reversible, and ATP-competitive p38 MAP kinase inhibitor (IC50 = 0.38 µM for p38α). Shown to effectively suppresses LPS-induced cytokine release both in vitro (IC50 = 0.16 and 0.039 µM for TNF-α and IL-1β release, respectively, in human PBMC) and in vivo (ED50 = 1.33 mg/kg for TNF-α release in mouse). When compared with SB 203580 (Cat. No. 559389 and 559398), it exhibits reduced inhibitory activity against cytochrome P450-2D6 isoform and, therefore, is better suited for in vivo use. A 10 mM (1 mg/247 µl) solution of p38 MAP Kinase Inhibitor III (Cat. No. 506148) in DMSO is also available.
A cell-permeable, potent, selective, reversible, and ATP-competitive inhibitor of p38 MAP kinase (IC50 = 380 nM for p38α). Shown to effectively suppress LPS-induced cytokine release in vitro (IC50 = 160 nM for TNF-α release and 39 nM for IL-1β release, human PBMC) and in vivo (ED50 = 1.33 mg/kg for TNF-α release in mice). Exhibits reduced inhibitory activity against cytochrome P450-2D6 compared to SB 203580 (Cat. No 559389) and is better suited for use in animal models.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
p38 MAP kinase
Product competes with ATP.
Reversible: yes
Target IC50: 0.38 µM for p38 MAP kinase α; 0.16 and 0.039 µM in suppressing LPS-induced TNF-α and IL-1β release, respectively, in human PBMC

Packaging

Packaged under inert gas

Warning

Toxicity: Carcinogenic / Teratogenic (D)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Laufer, S.A., et al. 2003. J. Med. Chem.46, 3230.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dai-Xu Li et al.
General physiology and biophysics, 39(6), 545-555 (2020-11-24)
Cardiovascular disease (CVD) states are associated with endothelial dysfunction (ED) and increased production of ROS in endothelial cells. The present study aimed to explore the protective effects of antioxidant protein peroxiredoxin 6 (PRDX6) on angiotensin II (AngII)‑induced human umbilical vein

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service