Skip to Content
Merck
All Photos(1)

Key Documents

506107

Sigma-Aldrich

Anti-P2X7 Receptor (576-595) Rabbit pAb

Sign Into View Organizational & Contract Pricing

Select a Size

1 SET
€322.00

€322.00


Estimated to ship on25 April 2025Details


Request a Bulk Order

Select a Size

Change View
1 SET
€322.00

About This Item

UNSPSC Code:
51111800
NACRES:
NA.41

€322.00


Estimated to ship on25 April 2025Details


Request a Bulk Order

Quality Level

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service