Skip to Content
Merck
All Photos(1)

Key Documents

5.05818

Sigma-Aldrich

Scopolamine Hydrobromide

Synonym(s):

Scopolamine Hydrobromide, Muscarinic Acetylcholine Receptor (mAChR) Antagonist, Scopolamine, Levo-duboisine, Hyoscine, Scopoderm

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€134.00

€134.00


Estimated to ship on26 May 2025Details


Request a Bulk Order

Select a Size

Change View
1 G
€134.00

About This Item

Empirical Formula (Hill Notation):
C17H21NO4 · xHBr
CAS Number:
Molecular Weight:
303.35 (free base basis)
MDL number:
UNSPSC Code:
12352116
NACRES:
NA.77

€134.00


Estimated to ship on26 May 2025Details


Request a Bulk Order

Assay

≥99% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

H2O: 100 mM

storage temp.

2-8°C

SMILES string

[Br].N1([C@@H]2[C@@H]3O[C@@H]3[C@@H]1CC(C2)OC(=O)[C@H](CO)c4ccccc4)C

InChI

1S/C17H21NO4.Br/c1-18-13-7-11(8-14(18)16-15(13)22-16)21-17(20)12(9-19)10-5-3-2-4-6-10;/h2-6,11-16,19H,7-9H2,1H3;/t11?,12-,13+,14+,15-,16+;/m1./s1

InChI key

XUZPYCBMTSSDHK-AKTDCHNFSA-N

General description

A competitive, non-selective antagonist for muscarinic acetylcholine receptors (mAChR). Most effective at treating motion sickness. It is also a research tool for inducing memory impairment in vivo. Frequently used in researches related to learning, memory and Alzheimer′s dementia.

Warning

Toxicity: Harmful (C)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Klinkenberg, I., et al. 2010. Neurosci Biobehav Rev34, 1307.
Aparkes. W. 1995. Psychopharmacologia.7, 1.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service