Skip to Content
Merck
All Photos(1)

Key Documents

5.04120

Sigma-Aldrich

Reboxetine Mesylate

Synonym(s):

Reboxetine Mesylate, (2R*)-2-[(R*)-(2-Ethoxyphenoxy)phenylmethyl]morpholine methanesulfonate, Edronax, Vestra

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€179.00

€179.00


Estimated to ship on02 June 2025Details


Request a Bulk Order

Select a Size

Change View
10 MG
€179.00

About This Item

Empirical Formula (Hill Notation):
C19H23NO3 · xCH4O3S
CAS Number:
Molecular Weight:
313.39 (free base basis)
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€179.00


Estimated to ship on02 June 2025Details


Request a Bulk Order

Assay

≥99% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles
protect from light

color

white

solubility

water: 20 mg/mL

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(O)C.N1C[C@@H](OCC1)[C@H](Oc3c(cccc3)OCC)c2ccccc2

InChI

1S/C19H23NO3.CH4O3S/c1-2-21-16-10-6-7-11-17(16)23-19(15-8-4-3-5-9-15)18-14-20-12-13-22-18;1-5(2,3)4/h3-11,18-20H,2,12-14H2,1H3;1H3,(H,2,3,4)/t18-,19-;/m1./s1

InChI key

CGTZMJIMMUNLQD-STYNFMPRSA-N

General description

A potent and selective inhibitor of noradrenalin uptake (Ki values are 1.1, 129 and >10000 nM for rat NET, SERT and DAT, respectively). Displays >1000-fold selectivity over α-adrenoceptors, 5-HT, dopamine and muscarinic ACh receptors. Orally active antidepressant.

Biochem/physiol Actions

Primary Target
Noradrenalin transporter
Target Ki: 1.1, 129 and >10000 nM for rat NET, SERT and DAT respectively

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze 9-20°C. Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Sacchetti, G., et al. 1999. Br. J. Pharmacol.128, 1332.

Wong, E., et al. 2000. Biol. Psychiatry.47, 818.
Millan, M., et al. 2001. J. Pharmacol. Exp. Ther.298,565.
Owen, J., et al. 2003 Neurosci. Lett.348, 171.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 1B

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service