Skip to Content
Merck
All Photos(1)

Documents

5015

Sigma-Aldrich

LIGHT DIAGNOSTICS PBS Packet & 100x Tween 20/azide

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116012
eCl@ss:
32160405

manufacturer/tradename

Chemicon®
Light Diagnostics

technique(s)

immunocytochemistry: suitable

shipped in

wet ice

General description

Our PBS packet and 100X Tween 20/azide package contains five packages of 10X PBS powder (each sufficient to make 1 L of buffer) and 5 vials of 100X Tween 20/azide solution (10 mL per vial). The product is designed to be rehydrated according to the label instructions and stored at 4°C. The product is designed to be used as wash, and the components can be mixed together to make a solution of PBS containing both Tween 20 and azide. It is best to mix the solutions together AFTER rehydration. The final concentration of the 1X Tween 20 is 0.05%, and the final concentration of the azide is 0.1%.

Application

Research Category
All

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

General Lab Use

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service