Skip to Content
Merck
All Photos(1)

Key Documents

5.00526

Sigma-Aldrich

NOX Inhibitor VII, Thr101

Synonym(s):

NOX Inhibitor VII, Thr101, NADPH Oxidase Inhibitor VII, 6-Fluoro-2- o-tolylbenzo[d]isothiazol-3(2H)-one, 6-Fluoro-2-(2-methylphenyl)-1,2-benzothiazol-3(2H)-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H10FNOS
CAS Number:
Molecular Weight:
259.30
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Product 5.00526 is not currently sold in your country. Contact Technical Service

Assay

≥98% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

light brown

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

SMILES string

Fc1cc2c(cc1)C(=O)N(S2)c3c(cccc3)C

InChI key

BUIZTXXZBDUOCR-UHFFFAOYSA-N

General description

A cell-permeable, selenium-to-sulfur substituted Ebselen derivative that exhibits similar NOX2 inhibitory activity as Ebselen by blocking p22phox c-terminal PRD (proline-rich domain) and p47phoxbis-SH3 interaction. Although less potent than Ebselen in cell-free p22phox PRD-p47phoxbis-SH3 binding (IC50 = 4 vs. 0.3 µM) and NOX2 activity (IC50 = 4 vs. 0.6 µM) assays, Thr101 is more potent and NOX2-selective than Ebselen in cell-based assays (Thr101 IC50 /Ebselen IC50 = 0.3/0.5, 3/0.15, 8/no inhibition, 8/0.7 µM, respectively, against NOX2, NOX1, NOX4, and NOX5) and exhibits no glutathione peroxidase activity.Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
blocks p22phox c-terminal PRD (proline-rich domain) and p47phoxbis-SH3 interaction

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Smith, S.M.E., et al. 2012. Chem. Biol.19, 752.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service