Skip to Content
Merck
All Photos(1)

Key Documents

499050

Sigma-Aldrich

Osteocalcin, Bovine Bone

Synonym(s):

Osteocalcin, Bovine Bone, Bone-Gla-Protein, BGP

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€478.00

€478.00


Estimated to ship on23 April 2025Details



Select a Size

Change View
100 μG
€478.00

About This Item

Empirical Formula (Hill Notation):
C263H372N66O82S2
Molecular Weight:
5834.29
UNSPSC Code:
12352202
NACRES:
NA.42

€478.00


Estimated to ship on23 April 2025Details


Assay

≥98% (SDS-PAGE)

Quality Level

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

pI 

4.0 - 4.5

shipped in

wet ice

storage temp.

−20°C

General description

Single chain vitamin K-dependent protein produced by osteoblasts. Osteocalcin purified from bovine bone. Binds to phospholipid vesicles in the presence of Ca2+ (Kd = 6 µM). Also binds hydroxylapatite. Possible role in bone mineralization, bone resorption, and bone formation. Note: Gla corresponds to γ-carboxyglutamic acid.

Warning

Toxicity: Standard Handling (A)

Sequence

H-Tyr-Leu-Asp-His-Trp-Leu-Gly-Ala-Pro-Ala-Pro-Tyr-Pro-Asp-Pro-Leu-Gla-Pro-Lys-Arg-Gla-Val-Cys²³-Gla-Leu-Asn-Pro-Asp-Cys²⁹-Asp-Glu-Leu-Ala-Asp-His-Ile-Gly-Phe-Gln-Glu-Ala-Tyr-Arg-Arg-Phe-Tyr-Gly-Pro-Val-OH (disulfide bond: 23 → 29)

Physical form

In 75 mM NaCl, 10 mM sodium phosphate buffer, pH 7.4.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

Carter, S.D., et al. 1996. J. Anim. Sci. 74, 2719.
Watson, K.E., et al. 1994. J. Clin. Invest.93, 2106.
Tracy, R.P., et al. 1990. J. Bone Min. Res.5, 451.
Gendreau, M.A., et al. 1989. J. Biol. Chem.264, 6972.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. Is the Osteocalcin, Bovine Bone #499050, uncarboxylated or decarboxylated?

    1 answer
    1. Osteocalcin, bovine (499050) undergoes carboxylation. A vitamin K dependent carboxylase is responsible for post-translational modification, resulting in the production of three g-carboxyglutamic acid residues at positions 17, 21 and 24. The mature protein consists of 49 amino acids and features a single intrachain disulfide bond connecting Cys23 to Cys29. Osteocalcin is derived from bovine bone.

      Helpful?

  2. Is the product O5761 - Osteocalcin Human uncarboxylated or decarboxylated?

    1 answer
    1. Osteocalcin, Human, with product number O5761, is carboxylated.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service