Skip to Content
Merck
All Photos(1)

Key Documents

487923

Sigma-Aldrich

Anti-Nitrotyrosine Mouse mAb (CC22.8C7.3)

liquid, clone CC22.8C7.3, Calbiochem®

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€320.00

€320.00


Estimated to ship on02 May 2025Details


Request a Bulk Order

Select a Size

Change View
50 μG
€320.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€320.00


Estimated to ship on02 May 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

CC22.8C7.3, monoclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity (predicted by homology)

all

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG2b

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NOS1(4842)

General description

Purified mouse monoclonal antibody. Recognizes nitrotyrosine-containing proteins.
Recognizes nitrotyrosine.
This Anti-Nitrotyrosine Mouse mAb (CC22.8C7.3) is validated for use in Immunoblotting for the detection of Nitrotyrosine.

Immunogen

peroxynitrite-treated KLH

Application

Immunoblotting (2 µg/ml)

Warning

Toxicity: Standard Handling (A)

Physical form

In PBS, 50% glycerol, 0.1% BSA, 0.02% sodium azide, pH 7.2.

Reconstitution

Reconstitute with H₂O to a final stock concentration of 1 mg/ml. Following reconsitution, aliquot and freeze (-20°C). Avoid freeze/thaw cycles of solutions.

Analysis Note

Positive Control
Mouse macrophages treated with peroxynitrite

Other Notes

Myatt, L., et al. 1996. Hypertension28, 488.
Beckman, J.S., et al. 1994. Biol. Chem. Hoppe-Seyler375, 81.
Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enrique Samper et al.
Free radical biology & medicine, 46(3), 387-396 (2008-11-29)
Lymphomas adapt to their environment by undergoing a complex series of biochemical changes that are currently not well understood. To better define these changes, we examined the gene expression and gene ontology profiles of thymic lymphomas from a commonly used
Teresa Tiago et al.
Biochemistry, 45(11), 3794-3804 (2006-03-15)
Exposure of myosin subfragment 1 (S1) to 3-morpholinosydnonimine (SIN-1) produced a time-dependent inhibition of the F-actin-stimulated S1 Mg(2+)-ATPase activity, reaching 50% inhibition with 46.7 +/- 8.3 microM SIN-1 for 8.7 microM S1, that is, at a SIN-1/S1 molar ratio of
Lalita Gupta et al.
Proceedings of the National Academy of Sciences of the United States of America, 102(11), 4010-4015 (2005-03-09)
In vivo responses of midgut epithelial cells to ookinete invasion of three different vector-parasite combinations, Aedes aegypti-Plasmodium gallinaceum, Anopheles stephensi-Plasmodium berghei, and A. stephensi-P. gallinaceum, were directly compared by using enzymatic markers and immunofluorescence stainings. Our studies indicate that, in
Sanjeev Kumar et al.
The Journal of biological chemistry, 279(51), 53475-53482 (2004-10-01)
Plasmodium berghei invasion of Anopheles stephensi midgut cells causes severe damage, induces expression of nitric-oxide synthase, and leads to apoptosis. The present study indicates that invasion results in tyrosine nitration, catalyzed as a two-step reaction in which nitric-oxide synthase induction
Seungho Choi et al.
Neuroscience, 384, 41-53 (2018-05-22)
Alzheimer's disease (AD) is the most common form of dementia that is often accompanied by mood and emotional disturbances and seizures. There is growing body of evidence that neurons expressing γ-aminobutyric acid (GABA) play an important role in regulation of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service