Skip to Content
Merck
All Photos(1)

Key Documents

48-625MAG

Millipore

MILLIPLEX® 2-Plex Phospo/Total mTOR - Cell Signaling Multiplex Assay

Sign Into View Organizational & Contract Pricing

Select a Size

96 EA
€1,300.00

€1,300.00


Product Availability: You will receive an order acknowledgement once the order has been entered in our system.

Request more information

Select a Size

Change View
96 EA
€1,300.00

About This Item

UNSPSC Code:
12161503
NACRES:
NA.47

€1,300.00


Product Availability: You will receive an order acknowledgement once the order has been entered in our system.

Request more information

species reactivity

human, rat, mouse

Quality Level

manufacturer/tradename

Milliplex®

technique(s)

multiplexing: suitable

detection method

fluorometric (Luminex® xMAP®)

storage temp.

2-8°C

General description

mTOR (mammalian target of rapamycin) is a Ser/Thr kinase belonging to the phosphatidylinositol 3-kinase-related kinase protein family. It mediates cell growth, cell proliferation, cell motility, cell survival, protein synthesis and transcription in response to growth factors and energy needs. mTOR is part of the Akt/mTOR pathway which is an important pathway that controls cellular metabolism. Dysfunction of this pathway has been implicated in various human diseases such as diabetes, obesity and cancer. mTOR is encoded by the MTOR gene. The mTOR catalytic subunit exists in two structurally distinct complexes: mTORC1 and mTORC2. Each of the two complexes are composed of different interacting proteins and localized to different subcellular compartments which affect its activation and function. The core components of mTORC1 is composed of mTOR, regulatory-associated protein of mTOR (Raptor) and mammalian lethal with SEC13 protein 8 (MLST8). The core components of mTORC2 is composed of mTOR, rapamycin-insensitive companion of mTOR (RICTOR), MLST8, and mammalian stress-activated protein kinase interacting protein 1 (mSIN1).The MILLIPLEX® MAP 2-Plex Phospho/Total mTOR kit contains all the components necessary to measure total mTOR and phosphorylated mTOR (Ser2448). The MILLIPLEX® MAP 2-Plex Phospho/Total mTOR magnetic bead kit has been developed for the simultaneous detection of phosphorylated mTOR (Ser2448) and total mTOR in a single well using the Luminex system. This detection assay is a rapid, convenient alternative to Western Blotting and immunoprecipitation procedures for the analysis of cell lysate samples..

Specificity

Cross-reactivity between the antibodies and any of the other analytes in this panel is non-detectable or negligible.

Application

The MILLIPLEX® 2-Plex Phospho/Total mTOR kit detects total mTOR and phosphorylated mTOR (Ser2448).An overnight (4°C) incubation is recommended for best results.This assay requires 25 μL diluted, filtered cell lysate per well.This kit must be run using the provided Assay Buffer.The suggested working range of protein concentration for the assay is 1 to 25 μg of total protein/well (25 μL/well at 40 to 1,000 μg/mL)Analytes available:mTOR (Total)mTOR (Ser2448)

Packaging

96-well plate

Storage and Stability

Recommended storage for kit components is 2 - 8°C.

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Uncover how cells communicate with MILLIPLEX® cell signaling multiplex assays. Multiplexing with cell signaling phosphoprotein assays based on Luminex® xMAP® technology helps researchers measure phosphoproteins and total proteins within the same or different pathways from a single sample.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service