Skip to Content
Merck
All Photos(1)

Key Documents

48-619MAG

Millipore

MILLIPLEX® Total/Phospho Erk - Cell Signaling Multiplex Assay

Sign Into View Organizational & Contract Pricing

Select a Size

96 EA
€1,280.00

€1,280.00


Product Availability: You will receive an order acknowledgement once the order has been entered in our system.

Request more information

Select a Size

Change View
96 EA
€1,280.00

About This Item

UNSPSC Code:
12161503
NACRES:
NA.47

€1,280.00


Product Availability: You will receive an order acknowledgement once the order has been entered in our system.

Request more information

species reactivity

human, mouse, rat

Quality Level

manufacturer/tradename

Milliplex®

technique(s)

multiplexing: suitable

detection method

fluorometric (Luminex® xMAP®)

storage temp.

2-8°C

General description

The MILLIPLEX® MAP Total/Phospho Akt (Catalog No. 48-618MAG) and Total/Phospho ERK (48-619MAG) Magnetic bead kits have been developed for the simultaneous detection of phosphorylated Akt (Ser473) and total Akt or phosphorylated ERK1/2 (Thr185/Tyr187) and total ERK in a single well using the Luminex system. These detection assays are rapid, convenient alternatives to Western Blotting and immunoprecipitation procedures for the analysis of cell lysate samples.

Specificity

Cross-reactivity between the antibodies and any of the other analytes in this panel is non-detectable or negligible.

Application

Intracellular Bead-Based Multiplex Assays using the Luminex technology enables the simultaneous relative quantitation of multiple phosphorylation or total pathway proteins in tissue and cell lysate samples.An overnight (4°C) incubation is recommended for best results.This assay requires 25 μL diluted cell lysate per well.This kit must be run using Assay Buffer 2 (provided).1 - 25 μg cell lysate/well (recommended starting concentration is 40 to 1,000 μg protein/mL)Analytes available:Erk (Total)Erk (Thr185/Tyr187)

Packaging

96-well plate

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Uncover how cells communicate with MILLIPLEX® cell signaling multiplex assays. Multiplexing with cell signaling phosphoprotein assays based on Luminex® xMAP® technology helps researchers measure phosphoproteins and total proteins within the same or different pathways from a single sample.

Questions

  1. Is it possible to combine the Total/Phospho 2-Plex Cell Signaling Kits including the following components: 48-618MAG - Akt, 48-631MAG - Akt1, 48-632MAG - Akt2, 48-633MAG - Akt3, 48-628MAG - CREB, 48-619MAG - Erk/MAPK1/2, 48-626MAG - IRS1, 48-622MAG - JNK, 48-625MAG - mTOR, 48-624MAG - p38, 48-623MAG - STAT3?

    1 answer
    1. The 2-plex Cell Signaling kits can be combined except for the Akt panels. The Akt Panels cannot be combined with another Akt panel but they can be combined with any other 2-plex kit.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service