Skip to Content
Merck
All Photos(1)

Key Documents

481971-M

Millipore

NADP, Monosodium Salt

A coenzyme that is widely distributed in living matter. Participates in oxidation-reduction reactions. Spectra (pH 7.0): 250 nm/260 nm: 0.78-0.88; 280 nm/260 nm: 0.16-0.26.

Synonym(s):

NADP, Monosodium Salt, β-Nicotinamide-adenine Dinucleotide Phosphate, TPN, Co II, Na

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€96.70
500 MG
€286.00
1 G
€430.00
5 G
€1,430.00

€96.70


Estimated to ship on07 April 2026Details


Request a Bulk Order

Select a Size

Change View
100 MG
€96.70
500 MG
€286.00
1 G
€430.00
5 G
€1,430.00

About This Item

Empirical Formula (Hill Notation):
C21H27N7O17P3 · Na
CAS Number:
Molecular Weight:
765.39
MDL number:
UNSPSC Code:
41106305
NACRES:
NA.28

€96.70


Estimated to ship on07 April 2026Details


Request a Bulk Order

Quality Level

description

Merck USA index - 14, 6348

Assay

≥92% (enzymatic)
≥95% (UV)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white to off-white

solubility

water: soluble

storage temp.

−20°C

SMILES string

[P](=O)(O[P](=O)(OCC3OC(C(C3O)O[P](=O)(O)O)[n]4c5ncnc(c5nc4)N)O)(OCC1OC(C(C1O)O)[n+]2cc(ccc2)C(=O)N)O

InChI

1S/C21H28N7O17P3/c22-17-12-19(25-7-24-17)28(8-26-12)21-16(44-46(33,34)35)14(30)11(43-21)6-41-48(38,39)45-47(36,37)40-5-10-13(29)15(31)20(42-10)27-3-1-2-9(4-27)18(23)32/h1-4,7-8,10-11,13-16,20-21,29-31H,5-6H2,(H7-,22,23,24,25,32,33,34,35,36,37,38,39)/p+1

InChI key

XJLXINKUBYWONI-UHFFFAOYSA-O

General description

A coenzyme that is widely distributed in living matter. Participates in oxidation-reduction reactions (spectral ratio: 0.78-0.88 at 250/260 nm, pH 7.0; 0.16-0.26 at 280/260 nm, pH 7.0).
Spectra (pH 7.0): 250 nm/260 nm: 0.78-0.88; 280 nm/260 nm: 0.16-0.26.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Avoid freeze/thaw cycles of solutions.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service