Skip to Content
Merck
All Photos(1)

Key Documents

481918

Millipore

Nicotinic Acid

A component of coenzymes NAD+ and NADP+.

Synonym(s):

Nicotinic Acid, Niacin, Pyridine-3-carboxylic Acid

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€52.20

€52.20


Estimated to ship on31 March 2026Details


Request a Bulk Order

Select a Size

Change View
100 G
€52.20

About This Item

Empirical Formula (Hill Notation):
C6H5NO2
CAS Number:
Molecular Weight:
123.11
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.21

€52.20


Estimated to ship on31 March 2026Details


Request a Bulk Order

Quality Level

Assay

≥99.5% (acidimetric)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

water: soluble

density

1.47 g/cm3 at 25 °C

cation traces

heavy metals (as Pb): ≤0.002%

shipped in

ambient

storage temp.

10-30°C

SMILES string

n1cc(ccc1)C(=O)[O-].[H+]

InChI

1S/C6H5NO2/c8-6(9)5-2-1-3-7-4-5/h1-4H,(H,8,9)

InChI key

PVNIIMVLHYAWGP-UHFFFAOYSA-N

General description

A component of coenzymes NAD+ and NADP+.
Precursor of the coenzymes NAD and NADP.

Warning

Toxicity: Irritant (B)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

379.4 °F - closed cup

Flash Point(C)

193 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service