Skip to Content
Merck

4.80934

SAFC

Hydrochloric acid

2 M, Emprove® EXPERT

Pharma Manufacturing

Synonym(s):

Hydrogen chloride solution

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
€62.00
4809349025
€342.00
4809349950
€8,268.00

€62.00


Estimated to ship on09 April 2025Details


Request a Bulk OrderRequest a Sample

Select a Size

Change View
1 L
€62.00
4809349025
€342.00
4809349950
€8,268.00

About This Item

UNSPSC Code:
12352323

€62.00


Estimated to ship on09 April 2025Details


Request a Bulk OrderRequest a Sample

Product Name

Hydrochloric acid 2 mol/L, EMPROVE® EXPERT

Quality Level

product line

EMPROVE® EXPERT

form

liquid

pH

<1 (20 °C in H2O)

density

1.03 g/cm3 at 20 °C

storage temp.

2-25°C

General description

EMPROVE® EXPERT

Analysis Note

Concentration (HCl)1.8 - 2.2 mol/lIdentitypasses testColor≤ 10 HazenAppearancepasses testFree Bromine or Free Chlorinepasses testFree chlorine (Cl)≤ 0.0001Sulfate (SO₄)≤ 0.0005 %Sulfite (SO₃)≤ 0.001 %Heavy metals (as Pb)≤ 0.0001 %As (Arsenic)≤ 0.00015 %Residual solvents (ICH Q3C)...excluded by production processResidue on ignition (as SO₄)≤ 0.008 %Evaporation residue≤ 0.010 %Endotoxins≤ 0.25 I.U./mlLotnumber (raw material)Filtered with 0.2 µm
Manufactured from Hydrochloric acid fuming 37% EMPROVE® ESSENTIAL PhEur, BP, JP, NF and purified water acc. PhEur, JP.
Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany

also commonly purchased with this product

Product No.
Description
Pricing

related product

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1


What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Material Qualification Dossier

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Operational Excellence Dossier

Available by subscription or for a fee

Operational Excellence Dossier

Available by subscription or for a fee

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service