Skip to Content
Merck
All Photos(1)

Key Documents

480457

Sigma-Aldrich

NF-κB Activation Inhibitor VII, CID-2858522

The NF-κB Activation Inhibitor VII, CID-2858522 controls the biological activity of NF-κB. This small molecule/inhibitor is primarily used for Inflammation/Immunology applications.

Synonym(s):

NF-κB Activation Inhibitor VII, CID-2858522, 1-(3,5-di-tert-butyl-4-hydroxyphenyl)-2-(2-(3-hydroxypropylamino)-5,6-dimethyl-1H-benzo[d]imidazol-1-yl)ethanone; CID-2858522; 2-aminobenzimidazole; Compound 1a

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€162.00

€162.00


Estimated to ship on02 May 2025Details



Select a Size

Change View
10 MG
€162.00

About This Item

Empirical Formula (Hill Notation):
C28H39N3O3
Molecular Weight:
465.63
UNSPSC Code:
12352200
NACRES:
NA.77

€162.00


Estimated to ship on02 May 2025Details


Quality Level

Assay

>95% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

off-white

solubility

DMSO: 100 mg/mL

shipped in

wet ice

storage temp.

−20°C

General description

A cell-permeable selective inhibitor of NF-κB activation induced by PKC pathway activators such as PMA (IC50 = 70 nM) in a HEK293 cell-based NF-κB luciferase reporter gene assay, by operating downstream of PKC but upstream of IKKβ, without inhibiting other NF-κB activation pathways. At 10 µM, this compound only inhibits Raf, TLK1, and JAK2 by 57%, 70%, and 53%, respectively, among a panel of 353 protein kinases. In addition, it attenuates CD3/CD28 and PMA/ionomycin-induced production of IL-2 by Jurkat T-cells (IC50<5 µM) and anti-IgM-stimulated proliferation of murine B-lymphocytes (IC50 = 2 µM).

Warning

Toxicity: Standard Handling (A)

Other Notes

Peddibhotla, S., et al. 2010. J. Med. Chem.53, 4793.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service