Skip to Content
Merck
All Photos(1)

Key Documents

480450

Sigma-Aldrich

NFF-2

A quenched fluorescence substrate for MMPs.

Synonym(s):

NFF-2, MCA-RPKPYANvaWMK(DNP)-NH₂

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€412.00

€412.00


In StockDetails


Request a Bulk Order

Select a Size

Change View
1 MG
€412.00

About This Item

Empirical Formula (Hill Notation):
C79H105N19O19S
Molecular Weight:
1656.86
UNSPSC Code:
12352200
NACRES:
NA.77

€412.00


In StockDetails


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated
protect from light

color

yellow

solubility

DMSO: 5 mg/mL

fluorescence

λex ~325 nm
λem ~393 nm

shipped in

ambient

storage temp.

−20°C

General description

A quenched fluorescence substrate for MMPs. MMP-3 (stromelysin-1) hydrolyzes this substrate (kcat/Km = 5.9 x 104 M-1sec-1) at a rate 60 times faster than the hydrolysis by MMP-1 (interstitial collagenase). NFF-2 shows little discrimination between MMP-3, MMP-2 (gelatinase-A; kcat/Km = 5 x 104 M-1s-1), and MMP-9 (gelatinase-B; kcat/Km = 5.5 x 104 M-1s-1).
A quenched fluorescent substrate for MMPs (excitation maximum: ~325 nm; emission maximum: ~393 nm). MMP-3 (stromelysin-1) hydrolyzes this substrate (kcat/Km = 59,400 M-1s-1) at a rate 60X faster than hydrolysis by MMP-1 (interstitial collagenase). NFF-2 shows little discrimination between MMP-3, MMP-2 (gelatinase-A; kcat/Km = 54,000 M-1s-1), and MMP-9 (gelatinase-B; kcat/Km = 55,300 M-1s-1).

Biochem/physiol Actions

Cell permeable: no
Primary Target
Matrix Metalloproteinases (MMPs)
Product does not compete with ATP.
Reversible: no
kcat/Km = 5.9 x 104 M-1sec-1 as substrates for MMP-3 (stromelysin-1)

Warning

Toxicity: Standard Handling (A)

Sequence

MCA-Arg-Pro-Lys-Pro-Tyr-Ala-Nva-Trp-Met-Lys(DNP)-NH₂ (DNP = 2,4-dinitrophenyl; Nva = L-norvaline)

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Nagase, H., et al. 1994. J. Biol. Chem. 269, 20952.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service