Skip to Content
Merck
All Photos(2)

Key Documents

4.80448

Supelco

Acetonitrile solution

HPLC grade, suitable for HPLC, LiChrosolv®, contains 0.1% (v/v) trifluoroacetic acid

Synonym(s):

Acetonitrile solution, ACN, ACN + TFA, Cyanomethane, Ethyl nitrile, Methyl cyanide

Sign Into View Organizational & Contract Pricing

Select a Size

2.5 L
€422.00
30 L
€3,150.00
185 L
€15,730.00

€422.00


Estimated to ship on31 March 2025Details


Request a Bulk Order

Select a Size

Change View
2.5 L
€422.00
30 L
€3,150.00
185 L
€15,730.00

About This Item

Linear Formula:
CH3CN
CAS Number:
Molecular Weight:
41.05
Beilstein:
4124158
MDL number:
UNSPSC Code:
12191502
NACRES:
NA.03
grade:
HPLC grade
technique(s):
HPLC: suitable
vapor pressure:
97 hPa ( 20 °C)

€422.00


Estimated to ship on31 March 2025Details


Request a Bulk Order

Product Name

Acetonitrile with 0.1% (v:v) trifluoroacetic acid, for liquid chromatography LiChrosolv®

grade

HPLC grade

Quality Level

vapor pressure

97 hPa ( 20 °C)

description

isocratic

product line

LiChrosolv®

form

liquid

concentration

0.095-0.105 % (v/v) (trifluoroacetic acid)

technique(s)

HPLC: suitable

transition temp

flash point 2 °C (Acetonitrile)

density

0.782 g/mL at 25 °C

format

mixture

storage temp.

2-30°C

SMILES string

CC#N

InChI

1S/C2H3N/c1-2-3/h1H3

InChI key

WEVYAHXRMPXWCK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

With their high degree of UV transmittance, low particle count, low acidity and alkalinity and low evaporation residue level, LiChrosolv solvents are ideal for reproducible separations. Since separations are normally carried out under gradient conditions in analytical HPLC, we offer solvents in "gradient grade" as well as "isocratic grade". This enables to minimize the gradient effect of the solvent involved.

Application


  • Retention time prediction for post-translationally modified peptides: Ser, Thr, Tyr-phosphorylation.: This study presents a methodology for predicting retention times of phosphorylated peptides using high-performance liquid chromatography (HPLC) (Battellino et al., 2024).

  • Development and validation of a UPLC-PDA method for quantifying ceftazidime in dried blood spots.: This paper outlines a validated ultra-performance liquid chromatography photodiode array (UPLC-PDA) method for the quantification of ceftazidime, demonstrating its application and accuracy in analyzing dried blood samples (Lv et al., 2024).

  • A facile fabric phase sorptive extraction method for monitoring chloramphenicol residues in milk samples.: Describes a new method using fabric phase sorptive extraction for the efficient monitoring of chloramphenicol in milk, highlighting its simplicity and effectiveness (Ben Ayed et al., 2024).

  • Study of sample preparation influence on bacterial lipids profile in matrix-assisted laser desorption/ionization time-of-flight mass spectrometry.: This research investigates how different sample preparation methods affect the lipid profiles of bacteria analyzed by MALDI-TOF MS, providing insights into optimizing protocols for better accuracy (Arendowski et al., 2024).

  • Simple and robust LC-MS/MS method for quantification of colistin methanesulfonate and colistin in human plasma for therapeutic drug monitoring.: This article details a robust and straightforward liquid chromatography-tandem mass spectrometry (LC-MS/MS) method for the simultaneous determination of colistin methanesulfonate and colistin in human plasma, useful in clinical settings (Kim et al., 2023).

Analysis Note

Assay (trifluoroacetic acid): 0.095 - 0.105 % (v/v)
Lotnumber (trifluoroacetic acid):
Lotnumber (acetonitrile):
Required quantity (trifluoroacetic acid):
Required quantity (acetonitrile):

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

LICHROSOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - (Acetonitrile)

Flash Point(C)

2 °C - (Acetonitrile)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service